missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 5.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
£34.85 - £88.55
Specifications
Description | Accuracy ± 0.01 pH units |
---|---|
pH | 5.00 ±0.01 |
Physical Form | Liquid |
Packaging | HDPE Bottle GL45 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444549
|
Reagecon
105025 |
1 L |
£34.85
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454549
|
Reagecon
505025 |
5 L |
£88.55
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.01 pH units | |
5.00 ±0.01 | |
Liquid | |
HDPE Bottle GL45 |