Learn More
pH 5.5 Monobasic/Dibasic Potassium Phosphate Buffered Eluent in Accordance with United States Pharmacopoeia (USP), Reagecon
Easy-to-use, tested, certified, NIST traceable, and shelf-stable, buffered eluent.
Brand: Reagecon USP8905
Description
- Buffered Eluent for Chromatography, Buffered Eluent Mobile Phase
- Developed, formulated, and specified in accordance with United States pharmacopeia requirements
- NIST traceable
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
pH 5.5 ± 0.02 pH units | |
500 mL | |
HDPE Bottle GL45 | |
Mobile Phase Eluent |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
USP8921G1 | Certificate of Analysis | 28/10/2021 | USP8905 |