missing translation for 'onlineSavingsMsg'
Learn More

pH 7.4 Phosphate Buffer Solution according to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

Tested, certified, NIST traceable, and shelf-stable phosphate buffer solution.

Brand:  Reagecon 4004800

Product Code. 11424779

  • £52.10 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Buffer Solution according to European pharmacopoeia (EP)
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Accuracy ± 0.02 pH units
7.4 ±0.02
1 L
Product monographs
Liquid
HDPE Bottle GL45
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers