Learn More
pH 7.41 Buffer Solution at 25°C According to European Pharmacopoeia (EP), Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand:  Reagecon EP741
Code : A0
Additional Details : Weight : 1.00000kg
Product Code. 10708448
  • £73.70 / 500mL
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
Accuracy ± 0.01 pH units
Liquid
HDPE Bottle GL45
7.41 ±0.01
500 mL