missing translation for 'onlineSavingsMsg'
Learn More

pH 7 Buffer Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep PH-BUFF7-500

Additional Details : Weight : 0.50000kg

Product Code. 16321216

  • £63.30 / 500mL
Estimated Shipment: 19-08-2024
to see stock.

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
pH7 SI Units

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

pH 7 Buffer Standard
500 mL
Various
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Triple-leached LDPE bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only