missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 9 Buffer Standard, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| pH | 9 SI Units |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | pH 9 Buffer Standard |
| Concentration | 9 mol/L |
| For Use With (Application) | pH Calibration |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Standard |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition (by Analyte or Components) | Various |
Safety and Handling
Signal Word
- Danger
- Reproductive toxicity Category 1A
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : boric acid
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction