Learn More
pH Buffer pH 5, Fisher Chemical™
Click to view available options
Quantity:
5 L
Description
Mercury free buffers at 25°C (pH4 and pH10 are coloured) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
pH | 5 |
Quantity | 5 L |