missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Sachets, METTLER TOLEDO™

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.
£50.00 - £90.85
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||||
10431721
![]() |
Mettler Toledo
51302079 |
30 x 20 mL | 10.0 |
£90.85
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12683516
![]() |
Mettler Toledo
51302079 |
30 x 20 mL | 10.0 | N/A | N/A | N/A | |||||
12693516
![]() |
Mettler Toledo
51302080 |
30 x 20 mL | 4, 7, 10 |
£87.20
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Traceable to NIST primary standards
Specifications
Satchets |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pH Buffer Sachets, METTLER TOLEDO™