missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ pH Buffer Solution
Offers a range of pH buffers to care for your Jenway pH electrodes. Jenway™ ph Buffer Solution also offers a range of storage solutions and fill solutions tailored to your specific probe.
£43.30 - £49.50
Specifications
For Use With (Equipment) | pH Electrode |
---|---|
Physical Form | Liquid |
Quantity | 10 x 12 mL |
Packaging | 12mL Sachet |
Description
Offers a range of pH buffers to care for your Jenway™ pH electrodes. There is also a range of storage solutions and fill solutions tailored to your specific probe.Specifications
pH Electrode | |
Liquid | |
10 x 12 mL | |
12mL Sachet |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Jenway™ pH Buffer Solution