missing translation for 'onlineSavingsMsg'
Learn More

pH Buffers, Twin Neck, Fisherbrand™

Perform accurate pH calibrations with color-coded, twin neck Fisherbrand™ pH Buffer Bottles.

Brand:  Fisherbrand BS07/500ML

13.15 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 10287063

  • £16.45 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Perform accurate pH calibrations with color-coded, twin neck Fisherbrand™ pH Buffer Bottles.

Buffer Color Codes:

  • Pink = pH 4.01
  • Yellow = pH 7.00
  • Blue = pH 10.01

Available in 500 mL bottles

TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH calibration
7.0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    28

    Hours

    2

    Minutes

    11

    Seconds

    39
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH Buffers, Twin Neck, Fisherbrand™ > pH 7.0

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.