missing translation for 'onlineSavingsMsg'
Learn More

pH Buffers, Twin Neck, Fisherbrand™

Product Code. p-8993003 Shop All Fisherbrand Products
Click to view available options
pH:
4.01
7.0
10.01
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10741614

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Perform accurate pH calibrations with color-coded, twin neck Fisherbrand™ pH Buffer Bottles.

Buffer Color Codes:

  • Pink = pH 4.01
  • Yellow = pH 7.00
  • Blue = pH 10.01

Available in 500 mL bottles

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) pH calibration
pH 4.01
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.