Learn More
Hanna Instruments™ pH/Conductivity/TDS Tester
Designed for high accuracy measurements using one meter. Hannna Instruments™ pH/Conductivity/TDS Tester includes easy to read dual LCD display of pH/°C or TDS/°C. Includes 'HOLD' function and replaceable pH electrode.
Brand: Hanna Instruments™ HI98130
Code : 04
Additional Details : Weight : 0.15500kg
Description
- Combo testers designed for high accuracy measurements using one meter
- Easy to read dual LCD, pH/°C or TDS/°C
- Automatic temperature compensation
- Automatic calibration
- 'Hold' function
- Automatic shut off
- Replaceable pH electrode
Specifications
±0.05 | |
Automatic | |
0.01 | |
0°C to 60.0°C | |
163 x 40 x 26 mm | |
0.01ppt (g/L) | |
pH: Automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4% /°C | |
pH/Conductivity/TDS Tester | |
±2% |
±0.5°C | |
-2.00 to 16.00 | |
1 or 2 with 2 sets of standard buffers (pH 4.01, 7.01, 10.01 or pH 4.01, 6.86, 9.18) | |
0.1°C | |
0.00 to 10.00ppt (g/L) | |
-2.00 to 16.00 pH | |
1.5V | |
100 hr. |