missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Hard Case pH Electrode
Useful for measuring potential of hydrogen ions. Fisherbrand™ Hard Case pH Electrodes have a body manufactured with Teflon™.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Excellent for emulsions, solutions, TRIS buffers
- Electrolyte KCI 3M + AgCl
- Standard tip
- pH range of 0 to 14
- Temperature range of 0° to 80°C
Specifications
BNC | |
Molded Head and Cable | |
Sleeve | |
0°C to 80°C | |
12 mm |
pH Electrode | |
Ag/AgCl | |
0 to 14 | |
120 mm |