missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva pH Electrode
Provides temperature-compensated pH measurements. Cytiva pH Electrode has a rounded tip.
Brand: Cytiva 18-1111-26
Code : GX
Additional Details : Weight : 0.90000kg
Description
- Perfluororubber (FFKM), Glass and Titanium (Ti)
- The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules
- Exceptions include: 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
Specifications
UNF 10-32 2 B finger tights | |
0.088 mL | |
1:1 mixture of pH 4 buffer and 1 M KNO3 |
50 bar | |
0 to 14 | |
4.23 mm |