missing translation for 'onlineSavingsMsg'
Learn More

pH Electrode Storage Solution, Reagecon™
BIOPHARMA

Product Code. p-8014327 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 12890327

Brand: Reagecon ESS01

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.

The response, accuracy and lifespan of pH electrodes can be substantially improved through correct handling, cleaning and storage. Designed for routine equipment maintenance, these solutions are safe to use with any sensitive electrode. Electrode storage solutions are made from potassium chloride, free from silver ion, and are suitable for all Blueline electrodes.

Specifications

Composition Potassium Chloride solution, without Silver ion
Concentration 3 mol/L KCl
For Use With (Equipment) pH and Reference Electrodes, All Blueline Electrodes
Filling Solution Type Electrode Storage
Quantity 1000 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.