Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand:  Jenway™ 99960-42
293.55 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 13121734
  • £319.82 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Specifications
BNC
Refillable Environmental Combination with Bulb End
measurements at low temperatures or in solutions of low ionic strength, general purpose pH measurement and accuratae Q.C. or research work
Glass
0°C to 50°C
12 mm
12 dia. x 120 mm L
120 mm
0 to 11
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Jenway™ pH Electrodes > Electrode type: refillable environmental combination with bulb end; Connector: BNC; For measurements at low temperatures or in solutions of low ionic strength, general purpose pH measurement and accuratae Q.C. or research work