Learn More
Cytiva pH Flow Cell
Brand:  Cytiva 18-1112-92
Product Code. 11380232
  • £873.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
pH flow cell, including dummy electrode, with a round tip.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
pH Flow Cell
Perfluororubber (FFKM), Titanium (Ti), Polytetrafluoroethylene (PTFE)
pH flow cell, with dummy electrode, round tip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva pH Flow Cell > Max Flow Rate: 100mL/m in.