missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ pH Indicator Paper Refill

Product Code. 10323981 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
1 to 11
1 to 14
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 10323981

Brand: Macherey-Nagel™ 90202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides optimal quality and reliable readings. Macherey Nagel™ pH Indicator Paper Refills are supplied in plastic reels that ensure long-term stability and protection against many external influences.

  • Ready to use
  • Manufactured from high quality filter paper
  • Colors of the color scales are specially mixed to perfectly match the reaction color of the indicator papers

Specifications

pH Range 1 to 11
Format Refill
Quantity 3 Pack
Type pH Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.