missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ pH Indicator Paper Refill
Shop All Macherey-Nagel Products
Click to view available options
pH Range:
1 to 11
1 to 14
Unit Size:
Pack of 3
Description
- Ready to use
- Manufactured from high quality filter paper
- Colors of the color scales are specially mixed to perfectly match the reaction color of the indicator papers
Specifications
Specifications
| pH Range | 1 to 14 |
| Format | Refill |
| Quantity | 3 Pack |
| Type | pH Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction