Learn More
Fisherbrand™ pH Indicator Paper Roll
Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.
Brand:  Fisherbrand™
18.51 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.06300kg
 View more versions of this product
Product Code. 15950872
  • £30.25 / 4 units
In Stock
to see stock.
Add to basket
Description
Description
  • The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
  • Allows very simple pH measurement
  • Proven standard for many applications
Specifications
Specifications
6.4 to 8
pH testing
<6.4, 6.4, 6.6, 6.8, 7.0, 7.2, 7.4, 7.6, 7.8, 8.0, >8.0
pH Indicator Paper