Learn More
Fisherbrand™ pH Indicator Paper Roll
Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.
Brand: Fisherbrand™
14.40 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.05400kg
Description
- The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
- Allows very simple pH measurement
- Proven standard for many applications
Specifications
8 to 10 | |
pH testing | |
8.0, 8.2, 8.4, 8.7, 9.0, 9.2, 9.6, 10.0 | |
pH Indicator Paper |