Learn More
Fisherbrand™ pH Indicator Paper Roll
Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.
Brand: Fisherbrand™ 90207.204231.1
14.40 GBP valid until 2024-10-18
Use promo code "21642" to get your promotional price.
Description
- The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
- Allows very simple pH measurement
- Proven standard for many applications
Specifications
4 to 7 | |
pH testing | |
4.0, 4,3, 4,6, 4.9, 5.2, 5.5, 5.8, 6.1, 6.4, 6.7, 7.0 | |
pH Indicator Paper |