Learn More
Fisherbrand™ pH Indicator Paper Rolls
Provides rapid and precise pH readings. Fisherbrand™ pH Indicator Paper Rolls includesa color scale printed on the cover for convenience.
Brand: Fisherbrand™
Additional Details : Weight : 0.01504kg
Description
- Filter paper strips soaked with one or several coloured indicators of pH
- Economical selection of indicator papers cover the entire range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows colour changes in steps of pH2
Specifications
1 to 14 | |
Color scale printed on cover | |
5 m |
Refill | |
3/Pk. |