Learn More
Fisherbrand™ pH Indicator Paper Rolls
Provides rapid and precise pH readings. Fisherbrand™ pH Indicator Paper Rolls includesa color scale printed on the cover for convenience.
Brand:  Fisherbrand™
Additional Details : Weight : 0.07560kg
 View more versions of this product
Product Code. 15595669
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Filter paper strips soaked with one or several coloured indicators of pH
  • Economical selection of indicator papers cover the entire range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows colour changes in steps of pH2
Specifications
Specifications
0 to 11
Color scale printed on cover
5 m
Roll
4/Pk.