missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Indicator Paper Sticks
Alle ansehen Fisherbrand Produkte
Click to view available options
pH Range:
0 to 6
7 to 14
Packungsgröße:
Pack of 100
Beschreibung
85mm x 6mm sticks with different indicator papers sealed onto each strip. The pH is determined by comparison with a color chart supplied. The sticks are sufficiently long to avoid contact between the fingers and the test sample.
- Provides outstanding measurements safely over the designated pH range
- Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
- Indicator is covalently bonded on the pads and does not bleed into the sample – can be left in sample for an indefinite time without contamination
- Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
- Available in a flat box so they fit into your pocket
Spezifikation
Spezifikation
| pH Range | 0 to 6 |
| Includes | Colour comparison chart |
| Quantity | 100 Sticks |
| Type | pH Indicator Paper Stick |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur