Learn More
Fisherbrand™ pH Indicator Paper Sticks
Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don’t need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.
£15.69 - £56.35
Specifications
Includes | Colour comparison chart |
---|---|
Quantity | 100 Sticks |
Type | pH Indicator Paper Stick |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
10642751
|
Fisherbrand™
|
0 to 14 |
£40.14
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11966106
|
Fisherbrand™
|
0 to 6 |
£15.74
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10353641
|
Fisherbrand™
|
3.6 to 6.1 |
£38.80
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10333501
|
Fisherbrand™
|
4.5 to 10 |
£43.34
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11386454
|
Fisherbrand™
|
6 to 7.7 |
£56.35
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11996056
|
Fisherbrand™
|
7 to 14 |
£15.89
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11346454
|
Fisherbrand™
|
0.3 to 2.3 |
£40.30
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11316454
|
Fisherbrand™
|
2 to 9 |
£37.60
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11376454
|
Fisherbrand™
|
5.1 to 7.2 |
£37.60
Pack of 100 |
In Stock
Log in to see stock. |
|||||
15384184
|
Fisherbrand™
|
5.1 to 7.2 |
£15.69
100 units |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11396454
|
Fisherbrand™
|
7.5 to 9.5 |
£38.80
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11360395
|
Fisherbrand™
|
3.1 to 8.3 |
£15.69
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10070552
|
Fisherbrand™
|
6 to 10 |
£15.89
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don’t need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.
85mm x 6mm sticks with different indicator papers sealed onto each strip. The pH is determined by comparison with a color chart supplied. The sticks are sufficiently long to avoid contact between the fingers and the test sample.- Provides outstanding measurements safely over the designated pH range
- Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
- Indicator is covalently bonded on the pads and does not bleed into the sample – can be left in sample for an indefinite time without contamination
- Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
- Available in a flat box so they fit into your pocket
Specifications
Colour comparison chart | |
100 Sticks | |
pH Indicator Paper Stick |