missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Indicator Paper Strips
Dip the pH test paper into the solution to be tested and compare the colour with the colour chart provided. Fisherbrand™ pH Indicator Paper Strips provide quick and easy pH determination.
Brand: Fisherbrand™ PH114S
This item is not returnable.
View return policy
Description
- Includes color chart label for easy reference and pH level determination
- 200 strips
Specifications
0 to 14 | |
Color indicator chart | |
pH Test Strip |
Strip | |
200/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction