missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Meter SevenDirect SD20
GREENER_CHOICE

Product Code. 17561006 Shop All Mettler Toledo Products
Click to view available options
Type:
SevenDirect SD20 HA Kit
SevenDirect SD20 Kit
SevenDirect SD20 LowVol Kit
SevenDirect SD20 Organic Kit
SevenDirect SD20 Pure H2O Kit
SevenDirect SD20 Solids Kit
SevenDirect SD20 Stirrer Kit
SevenDirect SD20 Viscous Kit
SevenDirect SD20, Meter Only
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17561006

Brand: Mettler Toledo™ 30671559

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
TRUSTED_SUSTAINABILITY

Specifications

Includes Instrument, power s upply, protective cover, Get Started guide, electrode holder EasyPlace, appropriate electrode, calibration solutions
Accuracy (Temperature) ±0.1°C between 0°C and 100°C
Accuracy (mV) 0.1 mV
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Measurement Modes pH with mV and Temperature
Calibration Linear/Segmented
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) -2000 to 2000 mV
Resolution (mV) 1, 0.1 mV
Calibration Points Up to 5
Temperature Range (Metric) -30°C to +130°C
Temperature Range (English) -22°F to +266°F
Resolution (Temperature) 0.1°C
Display Type Color TFT
Backlight Option Yes
Memory 2000 Standard Measurements
Data Points 2000
Log Function Type Automatic, Manual
IP Rating IP54
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (Metric) 850 g
Weight (English) 1.9 lb.
Channels 1
Temperature Sensor InLab Solids-Pro ISM pH/Temperature Sensor
Type SevenDirect SD20 Solids Kit
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.