missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Meter SevenDirect SD20
GREENER_CHOICE

The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

Brand:  Mettler Toledo™ 30671560

1562.75 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 17571006

  • £1645.00 / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.1°C between 0°C and 100°C
0.002
0.001, 0.01, 0.1
1, 0.1 mV
-22°F to +266°F
TFT, 7 in. Color
IP54
850 g
1
SevenDirect SD20 Viscous Kit
0.1 mV
PC, Printer, USB Stick
-2000 to 2000 mV
-30°C to +130°C
0.1°C
2000 Standard Measurements
195 x 205 x 65 mm
1.9 lb.
InLab Viscous-Pro ISM pH/Temperature Sensor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ pH Meter SevenDirect SD20 > pH Meter; Type: SevenDirect SD20 Viscous Kit; Dimensions (L x W x H): 195 x 205 x 65 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.