missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ pH-Neutral Detergent

pH-neutral detergent - concentrated, for gentle remove of residual contaminants; 1 kg

Brand:  BINDER™ 1002-0016

Product Code. 15661546

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains surface-active substances, complexing agent, corrosion inhibitor
  • pH value ≈ 7.5
  • Density: 1.06 g/mL
Specifications

Specifications

pH-neutral detergent - concentrated, for gentle remove of residual contaminants; 1 kg
Detergent
1 kg
All BINDER Chambers
Gentle Removal of Residual Contaminants
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers