Learn More
Macherey-Nagel™ pH Paper
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
Brand: Macherey-Nagel™ 90205
This item is not returnable.
View return policy
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
ISO 9001 | |
Roll | |
1 Each |
0.5 to 5.5 | |
0.5, 1.0, 1.5, 2.0, 2.5, 3.0, 3.5, 4.0, 4.5, 5.0, 5.5 | |
pH Paper |