Learn More
Macherey-Nagel™ pH Paper
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
Brand: Macherey-Nagel™ 90213
This item is not returnable.
View return policy
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
ISO 9001 | |
Roll | |
1 Each |
9 to 13 | |
9.0, 9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0 | |
pH Paper |