Learn More
Macherey-Nagel™ pH Paper
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
Brand: Macherey-Nagel™ 90208
This item is not returnable.
View return policy
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
ISO 9001 | |
Roll | |
1 Each |
5.4 to 7 | |
<5.4, 5.4, 5.7, 6.0, 6.2, 6.4, 6.7, 7.0 >7.0 | |
pH Paper |