Learn More
Macherey-Nagel™ pH Paper
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
Brand: Macherey-Nagel™ 90211
This item is not returnable.
View return policy
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
ISO 9001 | |
Roll | |
1 Each |
7.2 to 9.7 | |
<7.2, 7.2, 7.5, 7.8, 8.1, 8.4, 8.7, 9.0, 9.3, 9.7, >9.7 | |
pH Paper |