missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System
Measure pH and pH & ORP/Redox quickly and easily
£279.00 - £378.00
Specifications
For Use With (Application) | Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications |
---|---|
For Use With (Equipment) | YSI model 556 handheld multi probe meter |
Includes | Storage bottle with storage solution |
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
11970115
|
YSI™
005565 |
pH and pH/ORP probe |
£378.00
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
11960115
|
YSI™
005564 |
pH Probe |
£279.00
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- User-replaceble sensor allows for quick sensor replacements if needed
- No tools necessary
Specifications
Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications | |
YSI model 556 handheld multi probe meter | |
Storage bottle with storage solution |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System