missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH-Fix Test Strips, 7.9-9.8

Provides outstanding measurements safely over the designated pH range. Fisherbrand™ pH-Fix Test Strips, 7.9-9.8 are sufficiently long to avoid contact between the fingers and the test sample.

Brand:  Fisherbrand™

Code : 43

Additional Details : Weight : 0.03060kg

Product Code. 11306464

  • £37.60 / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Available in a flat box so they fit into your pocket
  • Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
  • Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
  • Indicator is covalently bonded on the pads and does not bleed into the sample - can be left in the sample for an indefinite period of time without contamination
Specifications

Specifications

ISO 13485 certified
pH indicator
pH Test Strip
7.9 to 9.8
100 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers