Learn More
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper
Used for presumptive testing of suspicious stains. Phadebas™ Saliva Amylase-detection Forensic Paper is based on the Phadebas Amylase Test tablets but follows a separate method developed by the Forensic Biology labs.
Brand:  Magle Life Sciences™ 1314
 View more versions of this product
Product Code. 12895552
  • £524.00 / Pack of 12
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Pre-manufactured papers;
  • Are applicable directly from package;
  • Include Batch number and Expiry date;
  • Are easy to handle and use;
  • Have negligible batch variations;
  • Give no difference between labs and consequently have higher credibility as evidence material
Specifications
Specifications
300 x 195 mm