missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Gel SPS
Phago'Gel SPS is intended for the hygienic surgical disinfection of hands by friction as an alternative to simple or hygienic hand washing, in the absence of water, during serial contact, or before any aseptic gesture.
Brand: Christeyns™ C65056
View more versions of this product
This item is not returnable.
View return policy
Description
- High tolerance
- Fast to use
- Bactericidal, fungicidal, mycobactericidal and virucidal
- Fragrance-free, dye-free, phenoxyethanol-free and paraben-free
Specifications
Alcohol-based Hand Sanitizer Gel | |
Gel | |
1000 mL |
Colorless | |
12 Airless Bottles with 3 mL Pump |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 12LT X PHAGORUB GEL SPS BOTTLE Airless 3ML
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Ethanol, tetradecanol