missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Soft Disinfectant Detergent

Phago'Soft is a disinfectant detergent for all high surfaces, glass surfaces and screens. It is also compatible with stainless steel.

£48.90 - £69.63

Specifications

Color-coded No
Form Clear Liquid
pH 3 to 3.7
Scent Fresh Scent
Sterility Non-sterile
View More Specs

Products 2
Product Code Brand Product Type Size Price Quantity & Availability  
Product Code Brand Product Type Size Price Quantity & Availability  
17821135
View Documents
Christeyns™
C65004
Disinfectant Surface Spray 750 mL
£48.90
4.50 L
Please to purchase this item. Need a web account? Register with us today!
17831135
View Documents
Christeyns™
C65006
Disinfectant Solution 5 L
£69.63
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Ready to use
  • Eco-certified
  • Approved for food contact
  • Active substance (peracetic acid) guarantees broad-spectrum microbiological effectiveness while minimizing its environmental impact through its degradation into acetic acid, oxygen and water
Specifications

Specifications

No
3 to 3.7
Non-sterile
Clear Liquid
Fresh Scent
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Christeyns™ Phago'Soft Disinfectant Detergent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.