Learn More
Christeyns™ Phago'Solution
Product Code. p-200000179
Click to view available options
Volume (Metric):
500 mL
1 L
5 L
Unit Size:
10 L
12 L
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17850255
Brand: Christeyns™ C65023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Phago'Solution is intended for hygienic and surgical hand disinfection as an alternative to simple or hygienic hand washing in the absence of a water point, during serial contact, and before any aseptic gesture.
  • Antimicrobial activity in 30 sec.
  • Dermatologically tested
  • Promotes a high level of compliance
Specifications
Product Type Hand Disinfectant
Color Colorless
Form Liquid
Quantity 20 Bottles
Volume (Metric) 500 mL