missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Wipes DASR

Can be used for the rapid disinfection of small furniture, trolleys, exterior surfaces of non-invasive medical devices, and small surfaces close to the patient.

Brand:  Christeyns™ 460372

 View more versions of this product

Product Code. 17840265

This item is not returnable. View return policy

Description

Description

  • Can be used without rinsing on surfaces that may be in direct contact with foodstuffs (table, work surface, trays, meal trolleys, slicers, choppers, knives, etc.)
  • Can also be used on toys for complete disinfection
  • Will not leave marks on surfaces
  • Ready-to-use
  • Broad disinfection spectrum
  • Ecocertified impregnation product
  • Food contact compatible
Specifications

Specifications

Disinfectant Wipe
17 cm
12 cm
White
6.7 in.
4.7 in.
Plastic
200 Wipes/Pk., 10 Pk./Cs.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X2000 PHAGOWIPES DASR (10X200)

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, Quaternaryammonium compounds,propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers