Learn More
Christeyns™ Phago'Wipes Spores Wipes
Phago'Wipes Spores Wipes are bactericidal, fungicidal, mycobactericidal, virucidal and sporicidal with peracetic acid as their active ingredient.
Brand: Christeyns™ 60012
Additional Details : CAS Number : 7722-84-1 Weight : 2.05000kg
Description
- Disinfectant detergent wipes for all surfaces, including non-invasive medical devices (operating table, operating light, monitors, cables and connectors, stethoscopes, reflex hammers, blood pressure monitors)
- Can be used, with rinsing, on surfaces that may be in direct contact with foodstuffs
- Formulated from an active substance (peracetic acid) guaranteeing broad-spectrum microbiological efficacy while ensuring respect for the environment through its biodegradability into acetic acid, oxygen and water
- Ready-to-use
Specifications
Sanitizing Wipes | |
17 cm | |
12 cm | |
100 Wipes/Pk., 6 Pk./Cs. |
6.7 in. | |
4.7 in. | |
Plastic |
Safety and Handling
- PHAGO'SPORE
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrogen peroxide, propan-2-ol