Learn More
Tocris Bioscience™ Phalloidin-Janelia Fluor 646
Red fluorescent F-actin probe
Brand:  Tocris Bioscience™ 7201/12U
Additional Details : Weight : 0.00970kg
Product Code. 18799556
  • £334.76 / 12µg
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
Phalloidin-Janelia Fluor® 646 is a red fluorescent F-actin probe. Phalloidin-Janelia Fluor® 646 is composed of the F-actin probe, Phalloidin (Cat. No. 4535), conjugated to Janelia Fluor® 646 (Cat. No. 6148). It is suitable for use in confocal and super-resolution microscopy techniques such as dSTORM. Excitation/emission maximum λ = 646/664 nm.
Specifications
Specifications
12 μg
Janelia Fluor Dyes
C64H75N11O13SSi
90%