Learn More
Proteintech Phantom Dye UV 450 Viability Dye
Phantom Dye UV 450 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00003-100TESTS
Product Code. 17318252
  • £50.00 / 100 tests
Explore more special offers
Description
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye UV 450 is a water soluble UV fluorescent dye excited by the UV laser (355 nm) making it an excellent addition to your multicolor panel.
Specifications
Specifications
Flow Cytometry
450 Emission
100 Tests