Learn More
Proteintech Phantom Dye Violet 540 Viability Dye
Phantom Dye Violet 540 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand:  Proteintech PD00006-100TESTS
Product Code. 17348252
  • £50.00 / 100 tests
Description
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Violet 540 is a water soluble violet fluorescent dye excited by the Violet laser (405 nm), emitting at 540 nm, making it an excellent addition to your multicolor panel.