Learn More
Masterflex™ PharMed™ BPT Hoses
Use this tubing for peristaltic pumps and sensitive cell cultures. Saint Gobain Performance Plastics™ PharMed™ BPT Hoses are less permeable to gases and vapors than silicone tubing and are ideal for fermentation, separation, purification and process monitoring and control systems.
Brand:  Masterflex™ 96880-08
Product Code. 15134663
  • £214.00 / 25ft
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Low particulate spallation
  • Can be autoclaved repeatedly
  • Withstands repeated CIP and SIP cleaning and sterilization
  • Fully tested to ISO 10993 standards to facilitate validation process
  • Meets USP Class VI, FDA and NSF criteria
Typical Applications
  • Diagnostic test product manufacturing
  • Cell harvest and media process systems
  • Vaccine manufacturing
  • Bioreactor process lines
  • Production filtration and fermentation
  • Sterile filling
  • Shear-sensitive fluid transfer
Specifications
Specifications
FDA ( 21 CFR 177.2600 ), NSF ( Standard 51 ), USP 23 Class VI made ​​
4.8 mm
7.5 m
Pharmaceuticals, Biotechnology, Transfer of cell cultures, cells or lichtempfind union media
1 Each
Opaque
135°C
Autoclavable
8.0 mm
1.6 mm
-50°C
Sterile
Flexible