missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences PharMed™ BPT Tubing

PharMed™ BPT Tubing is biocompatible tubing for peristaltic pumps and sensitive cell cultures.

Brand:  Saint Gobain Life Sciences R6502-26BPT

 View more versions of this product

Product Code. 10184234

  • £171.00 / 7.50m
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Low particulate spallation
  • Can be autoclaved repeatedly
  • Withstands repeated CIP and SIP cleaning and sterilization
  • Fully tested to ISO 10993 standards to facilitate validation process
  • Meets USP Class VI, FDA and NSF criteria
Typical Applications
  • Diagnostic test product manufacturing
  • Cell harvest and media process systems
  • Vaccine manufacturing
  • Bioreactor process lines
  • Production filtration and fermentation
  • Sterile filling
  • Shear-sensitive fluid transfer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FDA (21 CFR 177.2600), NSF (Standard 51), USP 23 Class VI Made
Autoclavable
1.6 mm
0.19 in.
7.5 m
Pharmaceuticals, Biotechnology, Transfer of cell cultures, cells or lichtempfind union media
1 Ea.
Opaque
135°C
Tubing
0.06 in.
PharMed BPT
4.8 mm
1.6 mm
-50°C
Sterile
Flexible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers