missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Pharmed Extension Tubing

For use in the pharmaceutical and food industries

Brand:  Gilson™ F1825124

 View more versions of this product

Product Code. 10404142

  • £158.00 / 3m
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
  • PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications

Specifications

Extension Pump Tubing
3.2 mm
300 cm
Pharmaceutical and food
1 Each
Low
135°C
Autoclavable
PharMed
For MF1 and MF4 Pump Heads
-60°C
77mL/min.
Opaque Beige
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers