missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ ThermoSafe™ Phase Change Soft Cold Packs

Use these cold packs for products requiring an accurate, consistent temperature. Cole-Parmer™ ThermoSafe™ Phase Soft Cold Packs are manufactured with laminated, metallized film exterior to resist punctures and provide a radiant heat barrier. Last fifteen percent longer than standard gel packs.

Brand:  Cole-Parmer™ 06347-74

 View more versions of this product

Product Code. 12328717

  • £68.65 / Pack of 24
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in 30 and °10°F (°1 and °23°C)  formulations.
  • High-tech phase change formula returns to gel state when thawed. 
  • Nontoxic and reusable
Specifications

Specifications

Phase Change Cold Pack
Laminated, Metallized Film
30°F
Keep sensitive materials at accurate and consistent temperature. Bottle capacity 354.9mL. Non-toxic and resuable
171 x 102 x 25 mm
Min. -1°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.