missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Phase Separating Paper, Grade: 902

Used for the separation of aqueous solutions from organic solutions

Brand:  Fisherbrand™ 0902A00005

116.39 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 11704043

  • £169.00 / Pack of 100
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Hydrophobic, with silicone impregnated paper
  • Organic phase permeates filter whereas aqueous phase, solids components are retained
  • Usable as disposable separating funnel
  • Density of organic phase has no influence on separating process
Specifications

Specifications

85 g/m2
Silicone
Hydrophobic
Phase Separating Paper
DIN 53137
Separating aqueous solutions from organic solutions, Usable as disposable separating funnel
0.17 mm
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers